Search
Search

UQCRC2| Gene Symbol: UQCRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945471.3 | XP_004945528.1 | 9 - 10 | 829 | 67 | |
| XM_424611.5 | XP_424611.2 | 9 - 10 | 832 | 67 | ||
| GenBank mRNA | BX950415.1 | - | 9 - 10 | 825 | 67 | |
Set Membership: |
|