Search
Search

FBXO38| Gene Symbol: FBXO38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154950.1 | XP_025010718.1 | 22 | 4091 | 128 | |
| XM_025154951.1 | XP_025010719.1 | 23 | 4117 | 128 | ||
| XM_025154952.1 | XP_025010720.1 | 23 | 4070 | 128 | ||
| XM_025154953.1 | XP_025010721.1 | 23 | 4158 | 128 | ||
| XM_025154954.1 | XP_025010722.1 | 22 | 3999 | 128 | ||
| XM_025154955.1 | XP_025010723.1 | 23 | 4082 | 128 | ||
| XM_025154956.1 | XP_025010724.1 | 23 | 4022 | 128 | ||
| XM_025154957.1 | XP_025010725.1 | 22 | 3990 | 128 | ||
Set Membership: |
|