Search
Search

PRDM16| Gene Symbol: PRDM16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947378.3 | XP_004947435.1 | 11 - 12 | 1473 | 65 | |
| XM_025142614.1 | XP_024998382.1 | 11 - 12 | 1473 | 65 | ||
| XM_025142615.1 | XP_024998383.1 | 11 - 12 | 1473 | 65 | ||
| XM_417551.6 | XP_417551.3 | 7 - 8 | 1091 | 65 | ||
Set Membership: |
|