Search
Search

ATP8B2| Gene Symbol: ATP8B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025143453.1 | XP_024999221.1 | 22 - 23 | 2816 | 92 | |
| XM_025143454.1 | XP_024999222.1 | 22 - 23 | 2832 | 92 | ||
| XM_025143455.1 | XP_024999223.1 | 19 - 20 | 2497 | 92 | ||
| XM_025143456.1 | XP_024999224.1 | 17 - 18 | 2330 | 92 | ||
Set Membership: |
|