Search
Search

C26H1orf106| Gene Symbol: C26H1orf106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298789.2 | XP_015154275.1 | 1285 | 131 | ||
| XM_015298790.2 | XP_015154276.1 | 1237 | 131 | |||
Set Membership: |
|