Search
Search

BCORL1| Gene Symbol: BCORL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278362.2 | XP_015133848.1 | 5 - 6 | 8197 | 93 | |
| XM_015278363.2 | XP_015133849.1 | 6 - 7 | 3220 | 93 | ||
| XM_025150323.1 | XP_025006091.1 | 5 - 6 | 3505 | 93 | ||
Set Membership: |
|