Search
Search

MICAL2| Gene Symbol: MICAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003641316.4 | XP_003641364.2 | 11 - 12 | 4012 | 76 | |
| XM_015286424.2 | XP_015141910.1 | 11 - 12 | 4057 | 76 | ||
| XM_015286425.2 | XP_015141911.1 | 12 - 13 | 4159 | 76 | ||
| XM_015286426.2 | XP_015141912.1 | 12 - 13 | 4114 | 76 | ||
| XM_015286428.1 | XP_015141914.1 | 12 - 13 | 1925 | 76 | ||
| XM_015286429.2 | XP_015141915.1 | 11 - 12 | 4012 | 76 | ||
| XM_015286430.2 | XP_015141916.1 | 11 - 12 | 4012 | 76 | ||
| XM_015286431.2 | XP_015141917.1 | 11 - 12 | 4011 | 76 | ||
| XM_015286432.2 | XP_015141918.1 | 11 - 12 | 4010 | 76 | ||
| XM_015286433.2 | XP_015141919.1 | 11 - 12 | 4004 | 76 | ||
| XM_015286434.2 | XP_015141920.1 | 11 - 12 | 4004 | 76 | ||
| XM_015286435.2 | XP_015141921.1 | 11 - 12 | 4004 | 76 | ||
Set Membership: |
|