Search
Search

ST18| Gene Symbol: ST18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015282653.2 | XP_015138139.2 | 9 - 10 | 1646 | 63 | |
| XM_025147958.1 | XP_025003726.1 | 10 - 11 | 2241 | 63 | ||
| XM_025147959.1 | XP_025003727.1 | 9 - 10 | 1784 | 63 | ||
| XM_025147960.1 | XP_025003728.1 | 9 - 10 | 1709 | 63 | ||
Set Membership: |
|