Search
Search

ITIH3| Gene Symbol: ITIH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154693.1 | XP_025010461.1 | 12 - 13 | 1695 | 129 | |
| XM_025154694.1 | XP_025010462.1 | 12 - 13 | 1542 | 129 | ||
| XM_414253.6 | XP_414253.4 | 13 - 14 | 1732 | 129 | ||
Set Membership: |
|