Search
Search

KLKB1| Gene Symbol: KLKB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025150183.1 | XP_025005951.1 | 9 - 10 | 1353 | 106 | |
| XM_025150184.1 | XP_025005952.1 | 9 - 10 | 1359 | 106 | ||
| XM_025150185.1 | XP_025005953.1 | 8 - 9 | 1296 | 106 | ||
| XM_025150186.1 | XP_025005954.1 | 8 - 9 | 1194 | 106 | ||
Set Membership: |
|