Search
Search

SMIM9| Gene Symbol: SMIM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940623.3 | XP_004940680.1 | 1 - 2 | 879 | 148 | |
| XM_015278396.2 | XP_015133882.1 | 1 - 2 | 879 | 148 | ||
Set Membership: |
|