Search
Search

BANK1| Gene Symbol: BANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015285617.2 | XP_015141103.2 | 13 - 14 | 2161 | 103 | |
| XM_420665.6 | XP_420665.6 | 13 - 14 | 2229 | 103 | ||
Set Membership: |
|