Search
Search

SLC4A11| Gene Symbol: SLC4A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015286072.2 | XP_015141558.2 | 12 - 13 | 1711 | 75 | |
| XM_025150089.1 | XP_025005857.1 | 11 - 12 | 1644 | 75 | ||
| XM_025150090.1 | XP_025005858.1 | 12 - 13 | 1447 | 75 | ||
| XM_025150091.1 | XP_025005859.1 | 12 - 13 | 1470 | 75 | ||
| XM_025150092.1 | XP_025005860.1 | 12 - 13 | 1826 | 75 | ||
| XM_025150093.1 | XP_025005861.1 | 11 - 12 | 1420 | 75 | ||
Set Membership: |
|