Search
Search

ECE2| Gene Symbol: ECE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943409.3 | XP_004943466.3 | 9 - 10 | 1249 | 78 | |
| XM_015291548.2 | XP_015147034.2 | 9 - 10 | 1249 | 78 | ||
| XM_025153673.1 | XP_025009441.1 | 9 - 10 | 1217 | 78 | ||
| XM_025153674.1 | XP_025009442.1 | 9 - 10 | 1249 | 78 | ||
Set Membership: |
|