Search
Search

ARAP2| Gene Symbol: ARAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936122.3 | XP_004936179.3 | 29 - 30 | 4718 | 94 | |
| XM_015285685.2 | XP_015141171.2 | 31 - 32 | 5140 | 94 | ||
| XM_426346.6 | XP_426346.6 | 29 - 30 | 4718 | 94 | ||
Set Membership: |
|