Search
Search

GPR156| Gene Symbol: GPR156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938173.2 | XP_004938230.1 | 9 - 10 | 1296 | 94 | |
| XM_025143459.1 | XP_024999227.1 | 8 - 9 | 1410 | 94 | ||
| XM_025143462.1 | XP_024999230.1 | 7 - 8 | 1315 | 94 | ||
| XM_025143472.1 | XP_024999240.1 | 7 - 8 | 1175 | 94 | ||
| XM_025143473.1 | XP_024999241.1 | 6 - 7 | 1079 | 94 | ||
| XM_416556.6 | XP_416556.3 | 9 - 10 | 2474 | 94 | ||
Set Membership: |
|