Search
Search

AIM1L| Gene Symbol: AIM1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947665.3 | XP_004947722.1 | 5 - 6 | 2717 | 75 | |
| XM_015297560.2 | XP_015153046.1 | 5 - 6 | 2715 | 75 | ||
Set Membership: |
|