Search
Search

PIEZO1| Gene Symbol: PIEZO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292664.2 | XP_015148150.1 | 38 - 39 | 5669 | 135 | |
| XM_015292666.2 | XP_015148152.1 | 38 - 39 | 5666 | 135 | ||
| XM_015292667.2 | XP_015148153.1 | 37 - 38 | 5597 | 135 | ||
Set Membership: |
|