Search
Search

KIF14| Gene Symbol: KIF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025153135.1 | XP_025008903.1 | 28 - 29 | 4585 | 111 | |
| XM_422190.6 | XP_422190.3 | 28 - 29 | 4584 | 111 | ||
Set Membership: |
|