Search
Search

RNF157| Gene Symbol: RNF157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015295309.2 | XP_015150795.1 | 6 - 7 | 720 | 97 | |
| XM_015295310.2 | XP_015150796.1 | 6 - 7 | 694 | 97 | ||
| XM_015295311.2 | XP_015150797.1 | 6 - 7 | 720 | 97 | ||
| XM_015295312.2 | XP_015150798.1 | 6 - 7 | 720 | 97 | ||
Set Membership: |
|