Search
Search

LOC548097
LOC100499653| Gene Symbol: LOC548097 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB086391.1 | - | 1 | 45 | 117 | |
| Gene Symbol: LOC100499653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250874.1 | NP_001237803.1 | 1 | 493 | 117 | |
| GenBank mRNA | FJ014870.1 | - | 1 | 493 | 117 | |
Set Membership: |
|