Search
Search

LOC547587| Gene Symbol: LOC547587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248057.1 | NP_001234986.1 | 1 - 2 | 360 | 66 | |
| GenBank mRNA | AF243374.1 | - | 1 - 2 | 341 | 66 | |
| BT090347.1 | - | 1 - 2 | 360 | 66 | ||
Set Membership: |
|