Search
Search

ACX1;2| Gene Symbol: ACX1;2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248635.1 | NP_001235564.1 | 11 - 12 | 1654 | 86 | |
| GenBank mRNA | AF404404.1 | - | 11 - 12 | 1654 | 86 | |
Set Membership: |
|