Search
Search

DR1| Gene Symbol: DR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248749.2 | NP_001235678.1 | 2 - 3 | 183 | 82 | |
| GenBank mRNA | AF464906.1 | - | 2 - 3 | 221 | 82 | |
| AK244706.1 | - | 2 - 3 | 224 | 82 | ||
| BT089816.1 | - | 2 - 3 | 185 | 82 | ||
| BT092715.1 | - | 2 - 3 | 227 | 82 | ||
Set Membership: |
|