Search
Search

AMS1| Gene Symbol: AMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249662.1 | NP_001236591.1 | 14 - 15 | 2059 | 76 | |
| GenBank mRNA | AK245291.1 | - | 14 - 15 | 2121 | 76 | |
| AY095999.1 | - | 14 - 15 | 2059 | 76 | ||
Set Membership: |
|