Search
Search

TP55
LOC100811186| Gene Symbol: TP55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249884.1 | NP_001236813.1 | 8 - 9 | 787 | 82 | |
| GenBank mRNA | AY250704.1 | - | 8 - 9 | 787 | 82 | |
| BT093713.1 | - | 8 - 9 | 772 | 82 | ||
| Gene Symbol: LOC100811186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK245949.1 | - | 8 - 9 | 794 | 82 | |
Set Membership: |
|