Search
Search

CDC48| Gene Symbol: CDC48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248170.1 | NP_001235099.1 | 8 - 9 | 2180 | 91 | |
| GenBank mRNA | AK286854.1 | - | 3525 | 91 | ||
| DQ359725.1 | - | 8 - 9 | 2180 | 91 | ||
| U20213.1 | - | 8 - 9 | 2200 | 91 | ||
Set Membership: |
|