Search
Search

LOC100037475| Gene Symbol: LOC100037475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249422.2 | NP_001236351.1 | 6 - 7 | 844 | 60 | |
| GenBank mRNA | AB196794.1 | - | 6 - 7 | 819 | 60 | |
| AK244547.1 | - | 6 - 7 | 821 | 60 | ||
Set Membership: |
|