Search
Search

LOC100037476| Gene Symbol: LOC100037476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249437.1 | NP_001236366.1 | 3 - 4 | 529 | 100 | |
| GenBank mRNA | AK287136.1 | - | 4 - 5 | 669 | 100 | |
| EF153101.1 | - | 3 - 4 | 529 | 100 | ||
Set Membership: |
|