Search
Search

WRKY10| Gene Symbol: WRKY10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250579.1 | NP_001237508.1 | 2 - 3 | 602 | 77 | |
| GenBank mRNA | BT093295.1 | - | 2 - 3 | 622 | 77 | |
| EU375344.1 | - | 2 - 3 | 602 | 77 | ||
Set Membership: |
|