Search
Search

LOC547950| Gene Symbol: LOC547950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249557.1 | NP_001236486.1 | 1 - 2 | 367 | 91 | |
| GenBank mRNA | AF243364.1 | - | 1 - 2 | 367 | 91 | |
| BT094624.1 | - | 1 - 2 | 91 | |||
Set Membership: |
|