Search
Search

LOC100500618| Gene Symbol: LOC100500618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249961.1 | NP_001236890.1 | 3 - 4 | 430 | 81 | |
| GenBank mRNA | AK245765.1 | - | 3 - 4 | 351 | 81 | |
| BT091558.1 | - | 3 - 4 | 430 | 81 | ||
Set Membership: |
|