Search
Search

LOC100305445| Gene Symbol: LOC100305445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248568.1 | NP_001235497.1 | 2 - 3 | 139 | 113 | |
| GenBank mRNA | FJ014832.1 | - | 2 - 3 | 139 | 113 | |
Set Membership: |
|