Search
Search

LOC100790835| Gene Symbol: LOC100790835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252978.1 | NP_001239907.1 | 2 - 3 | 181 | 89 | |
| GenBank mRNA | BT097027.1 | - | 2 - 3 | 181 | 89 | |
Set Membership: |
|