Search
Search

LOC100816028| Gene Symbol: LOC100816028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254152.1 | NP_001241081.1 | 3 - 4 | 300 | 115 | |
| GenBank mRNA | FJ014722.1 | - | 3 - 4 | 300 | 115 | |
Set Membership: |
|