Search
Search

LOC100799213| Gene Symbol: LOC100799213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254611.2 | NP_001241540.1 | 3 - 4 | 436 | 63 | |
| GenBank mRNA | BT094696.1 | - | 4 - 5 | 886 | 63 | |
Set Membership: |
|