Search
Search

LOC100803398| Gene Symbol: LOC100803398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255229.1 | NP_001242158.1 | 10 - 11 | 1220 | 66 | |
| GenBank mRNA | GQ422785.1 | - | 10 - 11 | 1220 | 66 | |
Set Membership: |
|