Search
Search

LOC100803398| Gene Symbol: LOC100803398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255229.1 | NP_001242158.1 | 11 - 12 | 1528 | 90 | |
| GenBank mRNA | GQ422785.1 | - | 11 - 12 | 1528 | 90 | |
Set Membership: |
|