Search
Search

LOC100783693| Gene Symbol: LOC100783693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255269.1 | NP_001242198.1 | 4 - 5 | 597 | 68 | |
| GenBank mRNA | GQ422782.1 | - | 4 - 5 | 597 | 68 | |
Set Membership: |
|