Search
Search

LOC100810354| Gene Symbol: LOC100810354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255406.2 | NP_001242335.1 | 2 - 3 | 490 | 99 | |
| GenBank mRNA | BT095483.1 | - | 2 - 3 | 492 | 99 | |
Set Membership: |
|