Search
Search

LOC100805314| Gene Symbol: LOC100805314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255730.2 | NP_001242659.1 | 2 - 3 | 481 | 101 | |
| GenBank mRNA | BT096023.1 | - | 2 - 3 | 483 | 101 | |
Set Membership: |
|