Search
Search

LOC100806373| Gene Symbol: LOC100806373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255865.2 | NP_001242794.1 | 6 - 7 | 1041 | 137 | |
| GenBank mRNA | BT093857.1 | - | 6 - 7 | 1042 | 137 | |
Set Membership: |
|