Search
Search

CYP74A1| Gene Symbol: CYP74A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289366.1 | NP_001276295.1 | 2 - 3 | 667 | 71 | |
| GenBank mRNA | AK245530.1 | - | 2 - 3 | 684 | 71 | |
| DQ340253.1 | - | 2 - 3 | 667 | 71 | ||
Set Membership: |
|