Search
Search

LOC100815232| Gene Symbol: LOC100815232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255455.2 | NP_001242384.1 | 3 - 4 | 466 | 79 | |
| GenBank mRNA | BT096112.1 | - | 3 - 4 | 468 | 79 | |
Set Membership: |
|