Search
Search

LOC100791193| Gene Symbol: LOC100791193 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255570.1 | NP_001242499.1 | 3 - 4 | 634 | 119 | |
| GenBank mRNA | BT093426.1 | - | 3 - 4 | 634 | 119 | |
Set Membership: |
|