Search
Search

LOC100809260| Gene Symbol: LOC100809260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252935.2 | NP_001239864.1 | 1 | 118 | 92 | |
| GenBank mRNA | BT099431.1 | - | 3 | 1236 | 92 | |
Set Membership: |
|