Search
Search

LOC100795221| Gene Symbol: LOC100795221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253160.2 | NP_001240089.1 | 5 | 1322 | 110 | |
| GenBank mRNA | BT096622.1 | - | 5 | 1324 | 110 | |
Set Membership: |
|