Search
Search

LOC100804022| Gene Symbol: LOC100804022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255690.2 | NP_001242619.1 | 11 | 1414 | 105 | |
| GenBank mRNA | BT093414.1 | - | 11 | 1416 | 105 | |
Set Membership: |
|