Search
Search

Bbs5| Gene Symbol: Bbs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857624.3 | XP_004857681.1 | 7 - 8 | 918 | 128 | |
| XM_004857626.2 | XP_004857683.1 | 7 - 8 | 652 | 128 | ||
| XM_021238626.1 | XP_021094285.1 | 7 - 8 | 745 | 128 | ||
Set Membership: |
|