Search
Search

Zmym2| Gene Symbol: Zmym2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854889.3 | XP_004854946.1 | 8 - 9 | 1947 | 103 | |
| XM_021236378.1 | XP_021092037.1 | 7 - 8 | 1875 | 103 | ||
| XM_021236379.1 | XP_021092038.1 | 9 - 10 | 1686 | 103 | ||
Set Membership: |
|